DuPont™ Tyvek™ IsoClean™ Series 264 Frocks, Clean-Processed and Sterile

Controlled environment processed to protect against particulates and other contaminants. Garments are gamma sterilized to SAL of 10-6. Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.

Manufacturer: DuPont™ IC264SWHMD0030CS

 View more versions of this product

Catalog No. 17-990-514

  Check Availability
Add to Cart

Alert

Should not be worn around open flame, heat source, sparks or ignition sources.


    Specifications

    Height (English) 5.3 to 5.5 ft.
    Length (English) Sleeve 35.5 in.
    Neck Style Bound
    Packaging Type Clean-processed
    Product Type Frock
    Closure Type Front Zipper
    Color White
    Cuff Style Elastic
    Length (English) 42.75 in.
    Material Flash-spun polyethylene
    Seam Type Serged
    Size Medium
    Sterile Yes,Yes
    Latex Free Yes
    Certifications/Compliance ISO Class 4 cleanroom (Former Fed. Std. 209 E Class 10)
    Height (Metric) 152.4 to 173.73cm
    For Use With (Application) Ideal balance of protection, durability, comfort and acts as barrier to dry particles, microorganisms and non-hazardous liquids
    Collar Type Bound neck
    Length (Metric) Sleeve 92.71cm
    Thickness 5mil
    Width (English) Chest 23.5 in.
    Width (Metric) Chest 59.69cm
    Size (English) Chest 32 to 36 in.
    Size (Metric) Chest 81.28 to 91.44cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit