DuPont™ Tyvek™ IsoClean™ Boot Covers

Excellent inherent barrier to dry particles, microorganisms and non-hazardous liquids

Manufacturer: Dupont Personal Protection IC457SWHMD01000B

 View more versions of this product

Catalog No. 17-988-089

  Check Availability
Add to Cart

    Specifications

    Material Tyvek™ Flashspun Polyethylene
    Height (Metric) 46.99cm
    Length (English) 12 in.
    Latex Free Yes
    Packaging Type Bulk packed
    Seam Type Serged
    Sterile No
    Color White
    Size Medium
    Sole Type PVC
    Product Type Boot Covers
    For Use With (Application) Ideal barrier to dry particles, microorganisms and nonhazardous liquids
    Height (English) 18.5 in.
    Closure Type Elastic top with ankle ties
    Length (Metric) 30.48cm
    Thickness 5.4mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit