Learn More
DuPont™ Tyvek™ IsoClean™ Coveralls
Create an excellent barrier to dry particles, microorganisms and nonhazardous liquids
Manufacturer:  Dupont Personal Protection IC258SWHXL00250B
 View more versions of this product
Catalog No. 17-988-053
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Specifications Description & Specifications
Specifications
Zipper
Elastic wrist and ankle, raglan sleeves, bound neck
White
40 to 43 in.
5.11 to 6.3 ft.
Excellent barrier to dry particles, microorganisms and non-hazardous liquids
99.69cm
88.9cm
5.4mil
Tyvek™ IsoClean™ Flashspun Polyethylene
Serged
X-Large
101.6 to 109.22cm
155.75 to 192.04cm
39.25 in.
35 in.
No