DuPont™ Tyvek™ IsoClean™ Coveralls

Create an excellent barrier to dry particles, microorganisms and nonhazardous liquids

Manufacturer: Dupont Personal Protection IC258SWHMD00250B

 View more versions of this product

Catalog No. 17-988-051

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


    Specifications

    Material Tyvek™ IsoClean™ Flashspun Polyethylene
    Seam Type Serged
    Color White
    Size Medium
    Height (Metric) 167.64 to 155.44cm
    For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids
    Length (English) Sleeve 36.75 in.
    Length (Metric) Sleeve 93.34cm
    Closure Type Zipper
    Size (English) Inseam 33 in.
    Size (Metric) Inseam 83.82cm
    Sterile No
    Thickness 5.4mil
    Includes Elastic wrist and ankle, raglan sleeves, bound neck
    Size (English) Chest 36 to 38 in.
    Size (Metric) Chest 91.44 to 96.52cm
    Height (English) 5.5 to 5.10 ft.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit