DuPont™ Tyvek™ IsoClean™ Coveralls

Create an excellent barrier to dry particles, microorganisms and nonhazardous liquids

Manufacturer: Dupont Personal Protection IC258SWH3X00250B

 View more versions of this product

Catalog No. 17-988-055

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


    Specifications

    Material Tyvek™ IsoClean™ Flashspun Polyethylene
    Includes Elastic wrist and ankle, raglan sleeves, bound neck
    Seam Type Serged
    Color White
    Size 3X-Large
    Size (English) Chest 46 to 52 in.
    Size (Metric) Chest 116.8 to 132.08cm
    Height (English) 6.2 to 6.6 ft.
    Height (Metric) 188.97 to 201.16cm
    For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids
    Length (English) Sleeve 42.5 in.
    Length (Metric) Sleeve 107.95cm
    Closure Type Zipper
    Size (English) Inseam 37 in.
    Size (Metric) Inseam 93.98cm
    Sterile No
    Thickness 5.4mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit