DuPont™ Tyvek™ IsoClean™ Coveralls

Create an excellent barrier to dry particles, microorganisms and nonhazardous liquids

Manufacturer: Dupont Personal Protection IC258SWH4X00250B

 View all options for this product

Catalog No. 17-988-056


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Closure Type Zipper
    Material Tyvek™ IsoClean™ Flashspun Polyethylene
    Includes Elastic wrist and ankle, raglan sleeves, bound neck
    Seam Type Serged
    Color White
    Size 4X-Large
    Size (English) Chest 52 to 58 in.
    Size (Metric) Chest 132.08 to 147.32cm
    Height (English) 6.3 to 7 ft.
    Height (Metric) 192.02 to 213.3cm
    For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids
    Length (English) Sleeve 44 in.
    Length (Metric) Sleeve 111.7cm
    Size (English) Inseam 38 in.
    Size (Metric) Inseam 96.52cm
    Sterile No
    Thickness 5.4mil

    • Made from polyolefin protective material
    • Comfortable, lightweight and durable
    • Bound neck with raglan sleeves
    • Serged seams
    • Elastic wrist and ankle
    • Zipper closure
    • Bulk packaged
    • White

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit