Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 105 White Coveralls
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Height (English):
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
Height (Metric):
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
Description
Light and durable coveralls provide coverage, including feet, while staying comfortable. Use in controlled environments with high standards for particle and microbiological contamination control.
- Clean processed and sterile
- Spinning process creates barrier to particles, micro-organisms, and non-hazardous liquids
- Lightweight, comfortable and durable
- Lowest level particle shedding
- Recyclable
- Serged seams
- Standard hood, elastic opening
- Elastic wrists and ankles
- Attached thumb loops
- Zipper closure
- Attached boots, PVC sole
Pharmaceutical, biotech, medical device, electronic settings
Specifications
Product Type | Cleanroom Coveralls |
Size | X-Large |
Material | Tyvek™ |
Color | White |
Ankle Style | Elastic, Attached Boots |
Cuff Style | Thumb Loop |
Neck Style | Hooded |
Hood Type | Elastic |
Closure Type | Zipper |
Seam Type | Serged |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction