DuPont ™ Tyvek ™ Isoclean ™ Series 105 White Coveralls

Excellent for a controlled environment to protect against particulates and other contaminants

Manufacturer: Dupont Personal Protection IC105SWH2X0025CS

 View more versions of this product

Catalog No. 17-685-213

  Check Availability
Add to Cart

    Specifications

    Material Tyvek™ IsoClean™ Flashspun Polyethylene
    Includes Elastic wrist and ankle, attached thumb loops, standard hood with elastic opening, attached boot with PVC soles, set sleeves
    Seam Type Serged
    Color White
    Size 2X-Large
    Size (English) Chest 43 to 46 in.
    Size (Metric) Chest 109.2 to 116.84cm
    Height (English) 6.1 to 6.5 ft.
    Height (Metric) 185.92 to 198.12cm
    For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids with lowest level of particle shedding within DuPont™ product portfolio
    Length (English) Sleeve 38.25 in.
    Length (Metric) Sleeve 97.15cm
    Closure Type Zipper
    Size (English) Inseam 30.5 in.
    Size (Metric) Inseam 77.4cm
    Sterile Yes
    Thickness 5mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit