Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont ™ Tyvek ™ Isoclean ™ Series 105 White Coveralls, Bulk Packaged

IsoClean is excellent for a controlled environment to protect against particulates and other contaminants

Manufacturer:  Dupont Personal Protection IC105SWHXL002500

 View more versions of this product

Catalog No. 17-685-202


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Zipper
Tyvek™ IsoClean™ Flashspun Polyethylene
101.6 to 109.22cm
Hooded
Serged
X-Large
155.75 to 192.04cm
Excellent barrier to dry particles, microorganisms and non-hazardous liquids
92.71cm
74.9cm
5.4mil
Elastic wrist and ankle, standard hood with elastic opening, set sleeves, attached thumb loops, attached boot with PVC soles
40 to 43 in.
Attached boots,Elastic
Elastic,Thumb loops
White
5.11 to 6.3 ft.
Elastic
36.5 in.
29.5 in.
Set
27.25 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit