Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ 224 Series Lab Coats
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
Light and durable labcoats provide upper body coverage, while staying comfortable.
- Spinning process creates barrier to particles, micro-organisms and nonhazardous liquids
- Lowest level particle shedding
- Serged seams
- Five-snap closure
- Laydown collar
- Two lower front pockets and one chest pocket
- Raglan sleeves
Applications:
Use in controlled environments with high standards for particle and microbiological contamination control; Pharmaceutical, biotech, medical device, electronic settings.
Specifications
Type | Lab Coat |
Size | Medium |
Material | Tyvek |
Color | White |
Closure Type | Snaps |
Cuff Style | Open |
Disposable | Single-use |
Sterility | Non-sterile |
No. of Pockets | 3 |
Collar Type | Lay Down |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction