Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Series 253 Coveralls, Sterile
BIOPHARMAGREENER_CHOICE

Catalog Number 17988039
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Sterile coveralls used in controlled environments to protect against particulates and other contaminants.

  • Garments are gamma sterilized to SAL of 10-6
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort
  • Full traceability on all sterilized apparel with certificates of sterility available
  • Garments are gamma sterilized to SAL of 10-6
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Coverall has elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Dolman sleeve design for greater range of motion and comfort
  • Full traceability on all sterilized apparel with certificates of sterility available
TRUSTED_SUSTAINABILITY
Product Type Cleanroom Coveralls
Size X-Large
Material Tyvek™
Color White
Ankle Style Covered Elastic
Cuff Style Covered Elastic
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Collar Type Bound
Model IC253BWH
Size (English) Chest 40 to 43 in.
Size (English) Inseam 33.5 in.
Size (Metric) Chest 101 to 109 cm
Size (Metric) Inseam 85 cm
Sleeve Style Dolman
Thickness 5 mil
For Use With (Application) Barrier to dry particles, microorganisms and non-hazardous liquids
Length (English) Sleeve 38.25 in.
Length (Metric) Sleeve 97.15 cm
Height (English) 5.11 to 6.3 ft.
Height (Metric) 155.75 to 192.04 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit