DuPont™ Tyvek™ IsoClean™ Series 253 Coveralls, Sterile

Excellent for a controlled environment to protect against particulates and other contaminants.

Manufacturer: DuPont™ IC253BWHMD00250S

 View more versions of this product

Catalog No. 17-988-037

  Check Availability
Add to Cart

Alert

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


    Specifications

    Material Tyvek™ IsoClean™ Flashspun Polyethylene
    Seam Type Bound
    Color White
    Size Medium
    Size (English) Chest 36 to 38 in.
    Height (English) 5.5 to 5.10 ft.
    Height (Metric) 167.64 to 155.44cm
    For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids
    Length (English) Sleeve 35.5 in.
    Length (Metric) Sleeve 90.17cm
    Closure Type Zipper
    Size (English) Inseam 30.5 in.
    Size (Metric) Inseam 77.4cm
    Sterile Yes
    Thickness 5mil
    Includes Covered elastic wrist and ankle, bound neck, dolman sleeves
    Size (Metric) Chest 91.44 to 96.52cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit