DuPont™ DuPont™ Tyvek™ IsoClean ™ Series 258 Coveralls

Protect against particulates and other contaminants in controlled environments

Manufacturer: DuPont™ IC258SWHLG00250C

 View more versions of this product

Catalog No. 17-988-058

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Material Tyvek™ IsoClean™ Flashspun Polyethylene
    Includes Raglan sleeves with bound neck, elastic wrist and ankle
    Seam Type Serged
    Color White
    Size Large
    Size (English) Chest 38 to 40 in.
    Size (Metric) Chest 96.52 to 101.6cm
    Height (English) 5.8 to 6 ft.
    Height (Metric) 176.78 to 182.88cm
    For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids
    Length (English) Sleeve 38 in.
    Length (Metric) Sleeve 96.52cm
    Closure Type Zipper
    Size (English) Inseam 34 in.
    Size (Metric) Inseam 86.36cm
    Sterile No
    Thickness 5mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit