Tyvek™ IsoClean ™ Series 253 Coveralls, Clean-Processed

Excellent for a controlled environment to protect against particulates and other contaminants.

Manufacturer: Dupont Personal Protection IC253BWHSM00250C

 View more versions of this product

Catalog No. 17-990-511

  Check Availability
Add to Cart

Alert

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


    Specifications

    Size Small
    Size (Metric) Chest 81.2 to 91.4cm
    Material Tyvek™ IsoClean™ Flashspun Polyethylene
    Includes Covered elastic wrist and ankle, bound neck, dolman sleeves
    Seam Type Bound
    Size (English) Chest 32 to 36 in.
    Height (English) 5 to 5.7 ft.
    For Use With (Application) Excellent barrier to dry particles, microorganisms and non-hazardous liquids with lowest level of particle shedding within DuPont™ product portfolio
    Length (English) Sleeve 35.75 in.
    Length (Metric) Sleeve 90.8cm
    Closure Type Zipper
    Size (Metric) Inseam 74.2cm
    Sterile No
    Thickness 5mil
    Color White
    Height (Metric) 152.4 to 173.7cm
    Size (English) Inseam 29.25 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit