Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Zipper Front Cleanroom Frocks, Raglan Sleeves

Catalog Number 17988080
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Length (English):
41.5 in.
43.75 in.
45.75 in.
47.75 in.
53.75 in.
55 in.
57 in.
Size (English) Chest:
28 to 32 in.
32 to 36 in.
36 to 38 in.
38 to 40 in.
40 to 43 in.
43 to 46 in.
46 to 52 in.
This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.
This item is not returnable. View return policy
Should not be worn around open flame, heat source, sparks or ignition sources.

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bulk packaged in double transparent poly liners.

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. ulk packaged in double transparent poly liners.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • A-line silhouette

 

TRUSTED_SUSTAINABILITY
Product Type Controlled Environments Frock
Size Small
Material Tyvek
Color White
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Seam Type Serged
Collar Type Bound
Size (English) Chest 28 to 32 in.
Size (Metric) Chest 71.12 to 81.28 cm
Sleeve Style Raglan Sleeve
Packaging Type Bulk Packaged
Length (English) 41.5 in.
Length (Metric) 105.41 cm
Length (English) Sleeve 35 in.
Length (Metric) Sleeve 88.9 cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit