DuPont™ Tyvek™ IsoClean™ Series 264 Frocks, Bulk

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. ulk packaged in double transparent poly liners.

Manufacturer: DuPont™ IC264SWHXL00300B

 View more versions of this product

Catalog No. 17-988-083

  Check Availability
Add to Cart

Alert

Should not be worn around open flame, heat source, sparks or ignition sources.


    Specifications

    Packaging Type Bulk
    Product Type Frock
    Closure Type Front Zipper
    Material Flash-spun polyethylene
    Sterile No
    Height (English) 5.8 to 6.1 ft.
    Height (Metric) 176.7 to 185.9cm
    Length (English) Sleeve 40 in.
    Length (Metric) Sleeve 101.6cm
    Neck Style Bound
    Size (English) Chest 38 to 40 in.
    Size (Metric) Chest 96.52 to 101.6cm
    Color White
    Cuff Style Elastic
    Length (English) 47.75 in.
    Length (Metric) 121.28cm
    Seam Type Serged
    Size X-Large

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit