Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 447 Boot Covers
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Length (Metric):
27.3 cm
33.7 cm
38.7 cm
43.8 cm
Sterility:
Non-sterile
Sterile
Description
Tyvek IsoClean fabric is antistatic and low linting and provides an excellent barrir to dry particulates, nonhazardous liquids and microorganisms.
- Coarsely textured interior to help prevent movement inside
- PVC Gripper soles provide excellent barrier to liquid penetration
- 18 in. (45.7cm) high Tyvek uppers with elastic leg and ankle openings
- Serged seams
- White
- Available clean processed and gamma sterilized or bulk packaged
Order Info
Clean and sterile covers are individually packaged in opaque bag and placed in double transparent poly liners in and ISO Class 4 cleanroom. Bulk covers are bulk packaged in double transparent poly liners.
Specifications
Size | X-Large |
Material | Tyvek Flashspun Polyethylene |
Latex Free | Yes |
Color | White |
Sterility | Non-sterile |
Closure Type | Elastic Top and Ankles |
Seam Type | Serged |
For Use With (Application) | Ideal to provide an excellent barrier to dry particles, nonhazardous liquids and microorganisms |
Length (English) | 15.25 in. |
Length (Metric) | 38.7 cm |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction