Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Series 451 Shoe Covers, Bulk

Light and durable shoe covers protect feet from contamination, while providing safety with Gripper™ sole

$138.00

Specifications

Closure Type Elastic
Color White
Material Tyvek™ Flashspun Polyethylene
Height (English) 5 in.
Height (Metric) 12.7cm
View More Specs

Products 3
Catalog Number Mfr. No. Size Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size Length (English) Length (Metric) Price Quantity & Availability  
17-988-087 Edge
View Documents
DuPont™
IC451SWHLG01000B
Large 14 in. 35.56cm
Case of 100 for $138.00
Edge
Add to cart
 
17-988-086
View Documents
DuPont™
IC451SWHMD01000B
Medium 11.75 in. 29.8cm
Case of 100 for $138.00
Add to cart
 
17-988-150
View Documents
DuPont™
IC451SWHXL01000B
X-Large 16 in. 40.64cm
Case of 100 for $138.00
Add to cart
 
Description

Description

Suitable for use in controlled environments with high standards for particle and microbiological contamination control (pharmaceutical, biotech, medical device and electronic settings).

  • Flashspun polyethylene creates barrier to particles, micro-organisms and non-hazardous liquids
  • Serged seams
  • Elastic opening
  • Gripper PVC soles

Should not be worn around open flame, heat source, sparks or ignition sources.

Specifications

Specifications

Elastic
Tyvek™ Flashspun Polyethylene
12.7cm
Serged
White
5 in.
Yes
Bulk packaged in double transparent poly liners
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit