DuPont™ Tyvek™ IsoClean™ Sterile Boot Covers

Create an excellent barrier to dry particles, microorganisms and nonhazardous liquids

Manufacturer: Dupont Personal Protection IC457SWHXL01000S

 View more versions of this product

Catalog No. 17-988-095

  Check Availability
Add to Cart

Alert

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


    Specifications

    Product Type Boot Covers
    Material Polyethylene
    Closure Type Elastic top with ankle ties
    Height (Metric) 46.99cm
    Length (English) 16.5 in.
    Latex Free Yes
    Seam Type Serged
    Color White
    Size X-Large
    Height (English) 18.5 in.
    Length (Metric) 41.91cm
    Sterile Yes
    Thickness 5.4mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit