Learn More
DuPont™ Tyvek™ IsoClean™ Sterile Boot Covers
Create an excellent barrier to dry particles, microorganisms and nonhazardous liquids
Manufacturer:  Dupont Personal Protection IC458BWHXL0100CS
 View more versions of this product
Catalog No. 19-813-220
  Check Availability
Add to cart
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Specifications Description & Specifications
Specifications
Elastic top with ankle ties
X-Large
Polyethylene
46.99cm
Yes
Bound
5.4mil
White
Gripper
18.5 in.
16.5 in.
41.91cm
Yes