DuPont™ Tyvek™ IsoClean™ Sterile Boot Covers

Create an excellent barrier to dry particles, microorganisms and nonhazardous liquids

Manufacturer: Dupont Personal Protection IC457SWHSM01000S

 View more versions of this product

Catalog No. 17-988-092

  Check Availability
Add to Cart

Alert

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.


    Specifications

    Size Small
    Product Type Boot Covers
    Material Polyethylene
    Height (English) 18.5 in.
    Closure Type Elastic top with ankle ties
    Height (Metric) 46.99cm
    Length (English) 10.5 in.
    Latex Free Yes
    Length (Metric) 26.67cm
    Seam Type Serged
    Sterile Yes
    Color White
    Thickness 5.4mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit