Learn More
DuPont™ Tyvek™ Plus 198 Series Coveralls
Tyvek Plus garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material that provides an ideal balance of protection, durability and comfort
Manufacturer:  Dupont Personal Protection TY198TWHMD002500
 View more versions of this product
Catalog No. 19-162-796
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Elastic
Serged and over-taped
Medium
Attached
36.25 in.
Flash-spun polyethylene
White
36 to 39 in.
Pharmaceutical manufacturing, medical applications, research and biosecurity laboratories, nuclear and maintenance
24