DuPont™ Tyvek™ Plus 198 Series Coveralls

Tyvek Plus garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material that provides an ideal balance of protection, durability and comfort

Manufacturer: Dupont Personal Protection TY198TWH2X002500

 View more versions of this product

Catalog No. 19-162-790

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Pharmaceutical manufacturing, medical applications, research and biosecurity laboratories, nuclear and maintenance
    Material Flash-spun polyethylene
    Seam Type Serged and over-taped
    Color White
    Size 2X-Large
    Size (English) Chest 46 to 49 in.
    Hood Type Attached
    Length (English) Sleeve 39.75 in.
    Width (English) Chest 30
    Ankle Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit