Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ DuPont™ Tyvek™ White Pants

Pants with open ankle and elastic at waist for easy donning and removal

$285.24

Specifications

Material Tyvek™
Color White
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-067-055
View Documents
DuPont™
TY350SWHSM005000
Small
Case of 50 for $285.24
Add to cart
 
19-067-054
View Documents
DuPont™
TY350SWHLG005000
Large
Case of 50 for $285.24
Add to cart
 
Description

Description

 Tyvek™ garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material. It provides an ideal balance of protection, durability and comfort of any limited use fabric technology.The fabric offers an inherent barrier against particles (down to 1.0 micron in size).
  • Suitable for applications that do not require full body coverage of coverall
  • Pants with elastic wrist, elastic waist and serged seams
  • Protection is built in fabric itself, there are no films or laminates to abrade or wear away
  • Fabric delivers consistently better barrier, even after wear and abrasion
Applications

Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up

Specifications

Specifications

Tyvek™
White
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit