Learn More
Bevco™ Dura Series Cleanroom ISO 4 Polyurethane Seating
Designed as the most durable polyurethane chair offering the greatest total body support. Advanced ergonomic features improve productivity and reduce fatigue for outstanding comfort and long-lasting use in the toughest work environments. Certified to meet ISO 4 specifications of ISO14644-1.
Manufacturer:  Bevco™ 7551DC
 View more versions of this product
Catalog No. 19-190-119
Add to cart
Description
Description
  • DURA Series Heavy Duty Cleanroom self-skinned polyurethane seating provides excellent cushioning properties while offering high-impact resistance for controlled environments.
    • These models are designed to meet the particulate goals of ISO 14644-1 with ISO 5 compliance certified by independent testing laboratory
    • Polyurethane seat and back are impervious to water, oil and most chemicals
    • Polyurethane texture is puncture resistant, easy-to-clean and provides non-slip surface
    • The generous seat size offers plenty of room for all-day comfort
    • The backrest provides ideal lumbar support to be highly effective throughout the day
    • Easy-to-use pneumatic height adjustment with lifetime cylinder guarantee
    • A high performance adjustable 20 in. chrome flat footring is standard on mid and tall height models
    • Adjustable armrests with polyurethane arm pads (available on tilt models only)
    • Mushroom glides standard
    • Dual-wheel hard floors casters are available (add two inches to seat height range)
    • 12 year warranty
    • Supports up to 350 lbs.
    • Meets ANSI/BIFMA standards
    • GreenGuard Gold certified
Specifications
Specifications
Seat Height, Tilt seat
Mushroom Glide
Polished Aluminum
Polyurethane
20.5 to 30.5 in.
Black
Chrome
Controlled Environment Chair
High Bench Height