Learn More
H-B Instrument™ Durac™ Alcohol Proof High Precision Hydrometers
Used during distilling and rectifying to measure percentage alcohol in various spirits
Manufacturer:  H-B Instrument™ B618076400
 View more versions of this product
Catalog No. 13-203-236
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5/Pk.
Lead free metal
129/140%
Alcohol Proof High Precision Hydrometers
0.1% Proof
368mm
15°C (60°F)