Learn More
H-B Instrument™ Durac™ Brix Sugar Scale Plain Form Hydrometers
Used in food and beverage and other industries for measuring percentage of sucrose by weight
Manufacturer:  H-B Instrument™ B618020400
 View more versions of this product
Catalog No. 13-203-225
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10/Pk.
Lead free metal
20°C (68°F)
Brix Sugar Scale Plain Form Hydrometer
±0.1° Brix
300mm
30/40° Brix