Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Durac™ Maximum-Registering Autoclavable Bimetal Thermometer: Without Individual Calibration Report Products for Science Education

Indicates the highest temperature achieved and hold that measurement until manually reset

Manufacturer:  H-B Instrument™ B613150000/EMD

Catalog No. S71880


Add to cart

Includes: Maximum-registering thermometer and multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy and instructions. 13-202-760 includes an individual Thermometer Calibration Report.

Description

Description

A maximum-registering pointer (red) is moved up by the temperature-measuring pointer (black) to indicate the highest temperature achieved

  • Threaded connection (1/4 in. NPT) screws directly into in-line processes or use to mount thermometer on included stand
  • Waterproof and pressurized
  • Withstands autoclave cycles
  • Constructed from durable 304 stainless steel with a glass lens
  • Withstands pressure, high temperatures and rust
  • Probe length: 50mm (2 in.)
  • Includes multilanguage (English, French, German, Italian, Portuguese and Spanish) instructions and Statement of Test and Calibration traceable to NIST™

Compliance

Traceable to NIST

Specifications

Specifications

−20°C
0°F
±1.5°C (±3°F) full range
0°F to 300°F
Traceable to NIST
81 mm
2 in.
Glass
150°C
300°F
2°C/5°F
-20°C to +150°C
3.19 in.
Without Individual Calibration Report
50 mm
Bimetal
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit