H-B Instrument™ Durac™ Specific Gravity ASTM Calibrated Hydrometers
Measures specific gravity of petroleum products and heavy liquids in accordance with American Society of Testing Materials (ASTM) E100 standards
Manufacturer: H-B Instrument™ B618981000
 View more versions of this product
Catalog No. 13-202-831
  Check Availability
Add to Cart
    Specifications
    Specific Gravity Range 0.750/0.800
    Temperature of Standardization 15°C (60°F)
    Product Type Specific Gravity ASTM Calibrated Hydrometers
    Quantity 10/Pk.
    ASTM Number 84H
    Ballast Type Lead free metal
    Length (Metric) 330mm
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback