Learn More
H-B Instrument™ Durac™ Specific Gravity and Baume Plain Form Hydrometers, For Liquids Heavier Than Water
Used in chemistry, pharmacology, food and beverage, and starch industries
Manufacturer:  H-B Instrument™ B618061200
 View more versions of this product
Catalog No. 13-203-233
Add to cart
Description
Description
  • Measure specific gravity/relative density (g/cm3) of liquids heavier than water and Baume
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization:15°C (60°F)
  • Hand inspected by our U.S. quality team
  • Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST
  • Universal model (13-202-121) is available for heavy and light liquids in a range of 0.700 to 2.00
Specifications
Specifications
10/Pk.
0.005
±0.5°
Specific Gravity and Baume Plain Form Hydrometers
±0.005
165mm
0.5°