H-B Instrument™ Durac™ Specific Gravity Precision Hydrometers
Used in food and beverage, chemical, pharmacology, transportation, refrigeration industries
Manufacturer: H-B Instrument™ B618011400
 View more versions of this product
Catalog No. 13-202-802
  Check Availability
Add to Cart
    Specifications
    Specific Gravity Range 1.420/1.490
    Temperature of Standardization 15°C (60°F)
    Product Type Specific Gravity Precision Hydrometer
    Accuracy 0.001 sp. gr.
    Ballast Type Lead free
    Length (Metric) 325mm
    Quantity 10/Pk.
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback