Learn More
H-B Instrument™ Durac™ Specific Gravity Precision Hydrometers
Used in food and beverage, chemical, pharmacology, transportation, refrigeration industries
Manufacturer:  H-B Instrument™ B618011700
 View more versions of this product
Catalog No. 13-202-805
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10/Pk.
Lead free
1.600/1.670
Specific Gravity Precision Hydrometer
0.001 sp. gr.
325mm
15°C (60°F)