Learn More
DWK Life Sciences DURAN™ Bottle Top Vacuum Filtration Units
Designed for sterilization or clarification of aqueous cell culture media.
Catalog Number | Mfr. No. | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Price | Quantity & Availability | |||||
14-209-161
|
DWK Life Sciences
292702826 |
0.2 μm | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Supplied as filter funnel only unit for use with DURAN TILT bottle (with GL 45 thread adaptor) or 45mm media bottles
- Available in three different asymmetric pore size (0.1, 0.2, or 0.45μm)
- Raised moulded graduation marks for easy reading of volume
- Manufactured in a Class 100,000 clean room from Class VI, non-cytotoxic materials
- Sterile
Specifications
Bottle Top Vacuum Filtration Unit |