Learn More
DWK Life Sciences DURAN™ Filtering Beakers with Heavy Wall
Manufacturer:  DWK Life Sciences 21 131 880 6
 View more versions of this product
Catalog No. 09-841-098
Add to cart
Description
Description
  • Easy-to-read scale and large labelling field for easy marking in fired-on, highly durable, white ceramic
  • Spout for clean pouring
Specifications
Specifications
390mm
Beaker
Borosilicate Glass
1/Pk.
15,000mL