Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences DURAN™ GL 45 Pouring Rings For Screw Caps
Click to view available options
Color:
Blue
Gray
Green
Yellow
Description
- Available in blue, green, yellow and grey
Specifications
Product Type | Pouring Rings For Screw Caps |
For Use With (Equipment) | Screw Cap |
Color | Blue |
Height (Metric) With Closure | 4mm |
Material | Polypropylene |
Autoclavable | Autoclavable |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction