Learn More
DWK Life Sciences DURAN™ GL 45 Protect Laboratory Glass Bottles W/o Screw-Cap and Pouring Ring
Manufacturer:  DWK Life Sciences 21 805 540 1
 View more versions of this product
Catalog No. 09-841-194
Add to cart
Description
Description
  • Choose bottle only or completed bottle with blue screw cap (PP1, integral lip seal) and pouring ring (PP1) for drip-free pouring and clean, safe working.
Specifications
Specifications
1000mL
GL 45 Glass Bottles
225mm
Plastic
101mm