DURAN™ Laboratory DG Safety Caps
screw cap GL 45 with one, two or three ports
Manufacturer: Wheaton Science Products 1168402
 View more versions of this product
Catalog No. 09-841-030
  Check Availability
Add to Cart
    Specifications
    Product Type Safety cap; 2 port
    For Use With Bottle
    Includes Connection screw, conical nipple, ventilation valve and membrane filter
    Neck Finish GL45
    No. per Case 1 Cs.
  • Description & Specifications
Related Products
Product Content Feedback