DURAN™ Laboratory DG Safety Waste Caps
additional connection option for activated carbon filter and leak hose
Manufacturer: Wheaton Science Products 1168418
 View more versions of this product
Catalog No. 09-841-036
  Check Availability
Add to Cart
    Specifications
    For Use With Premium bottle
    Includes Connection screw, conical nipple, ventilation valve and membrane filter
    Neck Finish GL45
  • Description & Specifications
Related Products
Product Content Feedback