Learn More
DWK Life Sciences DURAN™ Laboratory Glass Ceramic Lab Protection Plate
Manufacturer:  DWK Life Sciences 238215806
 View more versions of this product
Catalog No. 09-841-028
Add to cart
Description
Description
Laboratory mats Glass ceramic material with a smooth, pore-free surface.
Chemically resistant and operates between -200 and +700°C.
Ideal for use as a bench protector or heating surface in place of a wire gauze or pipeclay triangle.
Specifications
Specifications
Glass Ceramic
22cm
17.5cm
-328 and 1292°F
6.8 in.
8.66 in.
6.8 in.
Portection plate
-200 and 700°C
17.5cm