DURAN™ Laboratory Thread Adapters
Manufacturer: Wheaton Science Products 1168447
 View all options for this product
Catalog No. 09-841-045
  Check Availability
Add to Cart
    Product Type Thread Adapter
    For Use With (Application) Handling aggressive, gas generating media
    Material Polypropylene
    Thread Type GL38/GL45
    Adapters are available for GL 40, GL 38 and GL 32 screw threads.
  • Description & Specifications
Related Products
Product Content Feedback