DURAN™ Laboratory Thread Adapters
Manufacturer: Wheaton Science Products 1168449
 View more versions of this product
Catalog No. 09-841-047
  Check Availability
Add to Cart
    Specifications
    Product Type Thread Adapter
    For Use With (Application) Handling aggressive, gas generating media
    Material Polypropylene
    Thread Type GL32/GL45
  • Description & Specifications
Related Products
Product Content Feedback