Learn More
DWK Life Sciences DURAN™ Laboratory Thread Adapters
Manufacturer:  DWK Life Sciences 1168446
 View more versions of this product
Catalog No. 09-841-044
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Adapters are available for GL 40, GL 38 and GL 32 screw threads.
Specifications
Specifications
Thread Adapter
Handling aggressive, gas generating media
PTFE
GL38/GL45