Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ Square Laboratory Bottles with DIN Thread

Manufacturer:  DWK Life Sciences 21 820 365 5

 View more versions of this product

Catalog No. 09-841-286


Add to cart

Description

Description

  • Typical applications: space-saving storage, space-saving transport
  • With screw-cap and pouring ring from PP (blue)
  • Service temperature limit of screw cap and pouring ring: +140 °C
  • Easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable white ceramic
Specifications

Specifications

250mL
Square Laboratory Bottles
143mm
10/Pk.
64mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit