Learn More
Berkshire™ DURX™ 670 Wiper
Berkshire Durx™ 670 Wipers are smooth, highly absorbent and durable
Manufacturer:  Berkshire™ DR670202106
Catalog No. 19-066-727
Add to cart
Description
Description
  • Combination of synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Chemically compatible with common cleaning and disinfecting solutions
  • Autoclavable
  • Economical
  • Recommended for ISO Class 5 and above environments
Specifications
Specifications
Safety
21 x 20in.
21 in.
Cleanroom wipes
20in.
White
ISO Class 5 and above environments
Hydroentangled nonwoven blend: 55% Cellulose / 45% Polyester
150 sheets/pack, 6 packs/case