Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUS1L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548388

Catalog No. PA548388

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Catalyzes the synthesis of dihydrouridine, a modified base found in the D-loop of most tRNAs.
Specifications

Specifications

DUS1L
Polyclonal
PBS with 0.09% sodium azide
Q6P1R4
tRNA-dihydrouridine(16/17) synthase [NAD(P)(+)]-like, 131-, tRNA-dihydrouridine synthase 1-like, DUS1L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64118
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DUS1L
Liquid
DUS1L
KLH conjugated synthetic peptide between 321-349 amino acids from the C-terminal region of human DUS1L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit