Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUS2L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575960
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Dihydrouridine synthase catalyzes reduction of the 5,6-double bond of a uridine residue on the displacement loop of tRNA. The resultant modified base, 5,6-dihydrouridine, appears to increase the conformational flexibility and dynamic motion of tRNA (Kato et al., 2005 [PubMed 15994936]).
Specifications
DUS2L | |
Polyclonal | |
Unconjugated | |
DUS2 | |
2310016K04Rik; Dihydrouridine synthase 2; dihydrouridine synthase 2-like, SMM1 homolog; Dus2; Dus2l; hDUS2; RGD1305612; SMM1; SMM1 homolog; tRNA-dihydrouridine synthase 2-like; tRNA-dihydrouridine(20) synthase (NAD(P)(+)); tRNA-dihydrouridine(20) synthase [NAD(P)+]-like; Up-regulated in lung cancer protein 8; URLC8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
291978, 54920, 66369 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9D7B1, Q9NX74 | |
DUS2 | |
Synthetic peptide corresponding to amino acids 441-485 of Human DUS2L. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction