Learn More
DUS3L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 156431AP
Catalog No. 50-173-0371
Add to cart
Description
Description
DUS3L gene ontology annotations related to this gene include flavin adenine dinucleotide binding; metal ion binding; tRNA dihydrouridine synthase activity.
Specifications
Specifications
DUS3L
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3KRC5, Q91XI1, Q96G46
DUS3, DUS3L
Rabbit
IgG
150 μL
-20°C
Primary
224907, 301122, 56931
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DUS3L
Liquid
DUS3L
DUS3L Fusion Protein Ag8142
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat