Learn More
DUS4L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 218701AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-3618
Add to Cart
Description
Description
This antibody recognize the C-terminal of DUS4L.
Catalyzes the synthesis of dihydrouridine, a modified base found in the D-loop of most tRNAs.
Specifications
Specifications
DUS4L
Polyclonal
Unconjugated
DUS4L
DUS4, DUS4L, PP35
Rabbit
Antigen Affinity Chromatography
RUO
11062, 71916
-20°C
Liquid
Western Blot
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95620, Q32M08
Dus4l
DUS4L Fusion Protein Ag16145
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions