Learn More
Diversey™ Dusan™
Hard surface sanitizer
Manufacturer:  Diversey™ 1833470
 View more versions of this product
Catalog No. 17-000-055
Add to cart
Description
Description
  • Acidic low foaming iodophor
Specifications
Specifications
55 gal.
55 gal.
Brown
EPA, Kosher
Non-sterile
Dusan™
208 L
Drum
Sanitizing hard surfaces in the food service, healthcare and allied industries