Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUSP12 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259096

Catalog No. NBP259096


Add to cart

Description

Description

DUSP12 Polyclonal antibody specifically detects DUSP12 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

DUSP12
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
dual specificity phosphatase 12, dual specificity protein phosphatase 12, Dual specificity tyrosine phosphatase YVH1, DUSP1YVH1 protein-tyrosine phosphatase ortholog, EC 3.1.3.48, serine/threonine specific protein phosphatase, TDSP4, YVH1EC 3.1.3.16
Rabbit
IgG
100 ul
Protein Phosphatase
Primary
11266
Western Blot, Immunoprecipitation
Western Blot, Immunoprecipitation
Affinity Purified
DUSP12
Maps to a region between 1 and 50
Immunogen affinity purified
RUO
Store at 4C.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only